Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RC Mouse anti-Rat, PE, Clone: OX-22, BD

Catalog No. BDB554888 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554888 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554888 Supplier BD Biosciences Supplier No. 554888

Mouse Monoclonal Antibody

The OX-22 antibody reacts with CD45RC on pre-B lymphocytes, B cells, CD8+ T suppressor/cytotoxic cells, and a subset of CD4+ T helper (Th) lymphocytes. It weakly reacts with thymocytes. CD45RC is a high-molecular-weight isoform of CD45 (Leukocyte Common Antigen); its level of expression distinguishes subpopulations of CD4+ T cells with Th1-like and Th2-like effector functions. Levels of expression of CD45RC have also been reported to distinguish resting from activated T cells at various stages of maturation. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RC
Applications Flow Cytometry
Classification Monoclonal
Clone OX-22
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen PHA-activated rat lymph node cells
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.