Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RO Mouse anti-Human, APC, Clone: UCHL1, BD

Catalog No. BDB340438 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340438 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340438 Supplier BD Biosciences Supplier No. 340438
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45RO, a member of the CD45 family that includes CD45, CD45RA, and CD45RB, recognizes a 180-kilodalton (kd) isoform of the leucocyte common antigen (LCA). The CD45 antigen is a protein tyrosine phosphatase. In peripheral blood, the CD45RO antigen is present on approximately 40% of resting peripheral blood T lymphocytes, including the CD4+ and CD8+ subpopulations, as well as on most thymocytes and activated T lymphocytes. It is also expressed on monocytes, macrophages, and granulocytes. The CD45RO antigen is present at low density early in the T-lymphocyte maturation cycle. Upon activation by phytohemagglutinin (PHA) or alloantigen, naive T lymphocytes first acquire CD45RO and then lose CD45RA. When these activated T lymphocytes are rechallenged, the cells that exhibit a secondary response are primarily CD45RO+ , leading to the concept that CD45RO+ cells are a primed population of memory T lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RO
Applications Flow Cytometry
Classification Monoclonal
Clone UCHL1
Concentration 50μg/mL
Conjugate APC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human IL-2-dependent T-cell line
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.