Learn More
Description
Host Species: Mouse
Clone UCHL1
Isotype IgG2a, κ
Species Reactivity Hu
Immunogen Human IL-2-dependent T-cell line
Flow Cytometry
Specifications
Specifications
| Antigen | CD45RO |
| Classification | Monoclonal |
| Clone | UCHL1 |
| Conjugate | Brilliant Violet 510 |
| Host Species | Mouse |
| Immunogen | Human IL-2-dependent T-cell line |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.