Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD46 Clone: 8A9.1,

Catalog No. MABF293MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF293MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF293MI Supplier MilliporeSigma™ Supplier No. MABF293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD46 Clone: 8A9.1 in Human samples, and it is validated for Flow Cytometry, Immunocytochemistry, Western Blotting

Membrane cofactor protein (UniProt P15529; also known as Antigen identified by Monoclonal TRA-2-10, CD46 antigen, Complement membrane cofactor protein, Measles virus receptor membrane cofactor protein, TLX, Trophoblast leukocyte common antigen, Trophoblast-lymphocyte cross-reactive antigen) is encoded by the CD46 (also known as MCP, TLX, AHUS2, MIC10, TRA2.10) gene (Gene ID 4179) in human. Signal 1-34, Membrane cofactor protein (MCP) or CD46 is a widely expressed complement regulator that inhibits all three pathways of complement activation, classical (CP), alternative (AP), and lectin (LP). It binds to both C3b (AP) and C4b (CP) and acts as a cofactor for their inactivation by the C3b/C4b inactivator enzyme factor I. CD46 is initially produced with an N-terminal signal sequence (a.a. 1-34), which is post-translationally removed to yield the type 1 membrane bound CD46 consisting of a large extracellular (a.a. 35-343) portion, followed by a single transmembrane α-helical region (a.a. 344-366) and a cytoplasmic tail (a.a. 367-392) with several variant structures. The extracellular portion is composed of four extracellular Complement Control Protein (CCP) domains carrying the cofactor activity, a collar STP-rich region, and a short region of unknown function (U). CD46 also acts as a receptor for certain pathogens such as the measles virus (Edmonston strain), human herpesvirus 6, and Neisseria gonorrhoeae. CD46 is also implicated to have a role in human egg fertilization. Several mutations in the CCPs and the transmembrane region of CD46 are linked to atypical hemolytic uremic syndrome (aHUS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD46
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 8A9.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P15529
Gene Symbols CD46, MCP, MIC10
Host Species Mouse
Immunogen mIg Fc-tagged recombinant protein corresponding to the extracellular domain of human CD46.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_002380
Test Specificity Expected to react with isoforms A (no del, ABC1) and isoform B (del 13, ABC2). Reactivity toward other spliced isoforms (Uniprot P15529-3 through P15529-16) has not been determined.
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.