Learn More
Description
The B6H12 monoclonal antibody specifically binds to CD47, a 42-52 kDa N-linked glycan protein. CD47 antigen, also known as integrin-associated protein (IAP), is expressed on all hematopoietic cells, including leukocytes, platelets and erythrocytes. It is also expressed on epithelial cells, endothelial cells, fibroblasts and many tumor cell lines. CD47 may play a role as a signal transducer in the regulation of cation fluxes across cell membranes and in the chemotactic and adhesive interactions of leukocytes with endothelial cells. The B6H12 antibody is capable of blocking phagocytosis of microparticles by peripheral blood granulocytes. It has also been reported to induce proliferation of CD3-activated T cells.
Specifications
Specifications
| Antigen | CD47 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | B6H12 |
| Conjugate | Alexa Fluor 647 |
| Description | IAP; CDw149; gp42; MER6; Neurophilin; OA3 ; Rh-related antigen |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Purified human placental RGD-binding proteins |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.