Learn More
Description
Host Species: Mouse
Clone: B6H12
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Purified human placental RGD-binding proteins
Flow cytometry
Specifications
Specifications
| Antigen | CD47 |
| Classification | Monoclonal |
| Clone | B6H12 |
| Host Species | Mouse |
| Immunogen | Purified human placental RGD-binding proteins |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.