Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Mouse anti-Human, PE, Clone: B6H12, BD

Catalog No. BDB556046 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556046 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556046 Supplier BD Biosciences Supplier No. 556046
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The B6H12 monoclonal antibody specifically binds to CD47, a 42-52 kDa N-linked glycan protein. CD47 antigen, also known as integrin-associated protein (IAP), is expressed on all hematopoietic cells, including leukocytes, platelets and erythrocytes. It is also expressed on epithelial cells, endothelial cells, fibroblasts and many tumor cell lines. CD47 may play a role as a signal transducer in the regulation of cation fluxes across cell membranes and in the chemotactic and adhesive interactions of leukocytes with endothelial cells. The B6H12 antibody is capable of blocking phagocytosis of microparticles by peripheral blood granulocytes. It has also been reported to induce proliferation of CD3-activated T cells.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD47
Applications Flow Cytometry
Classification Monoclonal
Clone B6H12
Conjugate PE
Description IAP; CDw149; gp42; MER6; Neurophilin; OA3 ; Rh-related antigen
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified human placental RGD-binding proteins
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.