Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD48 Hamster anti-Mouse, Brilliant Violet 510, Clone: HM48-1, BD

Catalog No. BDB563536 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563536 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563536 Supplier BD Biosciences Supplier No. 563536
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The HM48-1 monoclonal antibody specifically binds to CD48 (previously known as BCM1 in mice, Blast-1 in human, and OX-45 in the rat), a GPI-anchored member of the Ig superfamily. It is widely distributed on leukocytes, but not on non-hematopoietic cells, and its ligands include CD2 (LFA-2) and CD244 (2B4 antigen). The HM48-1 mAb blocks binding of soluble CD2 to CD48-bearing cells, blocks the interaction of CD2 and CD244, inhibits spleen cell proliferative responses to mitogens, augments the proliferative response of spleen cells when cross-linked with anti-CD3e mAbs, and inhibits priming of CTL in vitro. In vivo administration of HM48-1 antibody can prolong survival of cardiac allografts, an effect which is greatly enhanced by the addition of anti-CD2 mAb 12-15. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leukocytes.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD48
Applications Flow Cytometry
Classification Monoclonal
Clone HM48-1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Description BLAST; BLAST-1; BCM1; HM48-1; MEM-102; Sgp-60; SLAMF2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse T lymphoma MBL-2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.