Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD48 Mouse anti-Human, FITC, Clone: TÜ145, BD

Catalog No. BDB555759 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555759 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555759 Supplier BD Biosciences Supplier No. 555759
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TÜ145 monoclonal antibody specifically binds to CD48. CD48 belongs to the CD2 family within the immunoglobulin superfamily. CD48 is a ∽43 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein. CD48 is strongly expressed on lymphocytes, monocytes and macrophages and weakly on granulocytes. The expression of CD48 is upregulated following lymphocyte activation and by Epstein-Barr virus (EBV) infection. CD48 binds with high affinity to CD244 and with low affinity to CD2. These receptor-ligand interactions can augment or inhibit leukocyte responses.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD48
Applications Flow Cytometry
Classification Monoclonal
Clone TÜ145
Conjugate FITC
Description BCM1; BLAST; BLAST1; Hulym3; MEM-102; OX45; SLAMF2; TCT.1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.