Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49a Hamster anti-Mouse, PE, Clone: Ha31/8, BD

Catalog No. BDB562115 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562115 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562115 Supplier BD Biosciences Supplier No. 562115
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The Ha31/8 monoclonal antibody specifically binds to the 180-kDa integrin α1 chain (CD49a), which is a transmembrane glycoprotein that non-covalently associates with the integrin β1 subunit (CD29) to form the α1β1 (complex known as VLA-1). VLA-1 has been reported to be expressed on activated T cells, monocytes, smooth muscle cells, and endothelial cells. It is a receptor for collagen and laminin. The Ha31/8 monoclonal antibody is specific for both rat and mouse CD49a. It has been reported that Ha31/8 antibody can block VLA-1-mediated binding of rat cells to collagen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49a
Applications Flow Cytometry
Classification Monoclonal
Clone Ha31/8
Concentration 0.2mg/mL
Conjugate PE
Description Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Rt Emulsified Lewis Rt Glomeruli Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.