Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49a Mouse anti-Human, PE, Clone: SR84, BD

Catalog No. BDB559596 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559596 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559596 Supplier BD Biosciences Supplier No. 559596
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a type I transmembrane molecule of approximately 200 kDa. CD49a is also known as very late antigen 1 α subunit (VLA-1) of the integrin family of cell adhesion molecules. CD49a associates with the integrin β 1 subunit (CD29) to form the α 1/ β 1 heterodimer (CD49a/CD29), and serves as receptor for collagen and laminin-1. It is expressed on activated T cells, monocytes, neuronal cells and smooth muscle cells. CD49a has been reported to play a role in cell attachment during the development of both the central and peripheral nervous systems.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49a
Applications Flow Cytometry
Classification Monoclonal
Clone SR84
Conjugate PE
Description Integrin α1 Chain
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen P-Selectin IgG1 Fusion Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.