Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49b Mouse anti-Human, Unlabeled, Clone: 2, BD

Catalog No. BDB611016 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611016 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611016 Supplier BD Biosciences Supplier No. 611016
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrins are a family of dimeric proteins that mediate cell-to-cell and extracellular matrix adhesion. They consist of a large α chain that is non-covalently associated with a smaller β chain which is used to define integrin subfamilies. These molecules exhibit a wide range of expression throughout development and adulthood. VLA-2 (very late antigen), a member of the integrin superfamily, was identified on activated T cells, but has since been reported to be on various cell types. VLA-2 is reported to be a heterodimer of integrin α2 (CD49b) and integrin β1 (CD29) subunits. The α2 chain contains a large extracellular domain, a transmembrane domain, and a short cytoplasmic tail. VLA-2 functions as a collagen receptor on platelets and fibroblasts, as well as a collagen and laminin receptor on endothelial and epithelial cells. On activated T cells, VLA-2, like LFA-1, exhibits increased number and affinity of ligand binding. Interactions of these molecules with their extracellular matrix ligands is important for directing effector T cells to their target tissues and to provide co-stimulatory signals. Thus, VLA-2 not only plays an important role in cellular adhesion, but may function in intracellular signal transmission.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49b
Applications Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Description Integrin α2; VLA-2α
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human VLA-2α aa.42-245
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.