Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49b Hamster anti-Rat, FITC, Clone: HA1/29, BD

Catalog No. BDB554999 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554999 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554999 Supplier BD Biosciences Supplier No. 554999
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The Ha1/29 antibody reacts with intergrin α2 chain (CD49b or VLA-2 α chain), the 160-kDa transmembrane glycoprotein that non-covalently associates with the intergrin β1 subunit (CD29) to form the intergrin α2β1 complex known as VLA-2. VLA-2, a receptor for collagen and laminin, is expressed on activated lymphocytes, epithelial cells, and platelets. It has been reported that Ha1/29 antibody can block VLA-2 binding to collagen and laminin. Ha1/29 antibody also reacts with mouse CD49b.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49b
Applications Flow Cytometry
Classification Monoclonal
Clone HA1/29
Concentration 0.5mg/mL
Conjugate FITC
Description Integrin α2 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Cultured Rat Glomerular Epithelial
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.