Learn More
CD49b Mouse anti-Human, PE-CF594, Clone: 12F1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564121
Description
The 12F1 monoclonal antibody specifically binds to the integrin α 2 chain (VLA- α 2), a member of the integrin family of extracellular matrix and cell-cell adhesion receptors. VLA- α 2 is non-covalently associated with VLA- β 1 chain (CD29) in the VLA-2 complex. α 2 is expressed on numerous cell types including activated T cells, platelets, and long-term cultivated T cells. α 2β
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

Specifications
CD49b | |
Monoclonal | |
PE-CF594 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
12F1 | |
Integrin α2 chain; ITGA2; VLA-2 alpha; Platelet GPIa; Collagen receptor; BR | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.