Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49c Mouse, Unlabeled, Clone: 42, BD

Catalog No. BDB611044 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611044 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611044 Supplier BD Biosciences Supplier No. 611044
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrins are a family of dimeric proteins that mediate cell-to-cell and extracellular matrix adhesion. They consist of a large α chain that is non-covalently associated with a smaller β chain which defines the integrin subfamilies. VLA-3 (Very Late Antigen-3), a member of the integrin superfamily, exhibits elevated expression on B lymphocytes, but is also found on monocytes, platelets, and hematopoietic progenitor cells. A heterodimer of α3 (CD49c) and β1 (CD29) subunits, VLA is a receptor for laminin, fibronectin, and collagen. The α3 chain contains a large extracellular domain with three putative metal-binding sequences, a transmembrane domain, and a short cytoplasmic tail. Differing requirements for divalent cations and the influence of RGD peptides results in multiple ligand-binding mechanisms for VLA-3. Although its expression is restricted in normal tissues, VLA-3 is found on a variety of cultured tumor cells. In addition, levels of VLA-3 have been shown to correlate with the degree of invasiveness of malignant melanoma cells. Thus, VLA-3 mediates intercellular adhesion and cell migration in normal and, possibly, cancerous cell types.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49c
Applications Western Blot
Classification Monoclonal
Clone 42
Concentration 250μg/mL
Conjugate Unconjugated
Description Integrin α3; VLA-3α
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse VLA-3α aa. 110-325
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Canine, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.