Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d Rat anti-Mouse, Unlabeled, Clone: 9C10 (MRF4.B), BD

Catalog No. BDB553314 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553314 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553314 Supplier BD Biosciences Supplier No. 553314
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 9C10 (MFR4.B) antibody reacts with the integrin α;4 chain (CD49d), which is expressed as a heterodimer with either of two β; chains, β;1 or β;7. The α;4β;1 integrin (VLA-4, CD49d/CD29) is expressed on most peripheral lymphocytes, thymocytes, and monocytes; while the α;4β;7 integrin (LPAM-1) is expressed on peripheral lymphocytes, but on only a small subset of thymocytes. These integrins mediate a variety of cell-cell and cell-matrix interactions, recognizing the ligands VCAM-1 (CD106) and fibronectin. Integrin α;4β;7 also preferentially binds to the mucosal vascular addressin, MAdCAM-1. Although the 9C10 (MFR4.B) antibody alone has been reported to have little function-blocking activity, it can augment the inhibitory effects of mAb R1-2 (Cat. No. 553153), resulting in almost complete inhibition of VLA-4 binding to VCAM-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49d
Applications Flow Cytometry
Classification Monoclonal
Clone 9C10 (MRF4.B)
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Integrin α4 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse (C57BL/6xA/J)F[1] Fetal Liver Mast MC/9
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.