Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d Rat anti-Mouse, PE, Clone: 9C10 (MRF4.B), BD

Catalog No. BDB557420 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557420 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557420 Supplier BD Biosciences Supplier No. 557420
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 9C10 (MFR4.B) monoclonal antibody specifically binds to the Integrin α 4 chain (CD49d), which is expressed as a heterodimer with either of two β chains, β 1 or β 7. The α 4β 1 integrin (VLA-4, CD49d/CD29) is expressed on most peripheral lymphocytes, thymocytes, and monocytes; while the α β 7 also preferentially binds to the mucosal vascular addressin, MAdCAM-1. Although the 9C10 (MFR4.B) antibody alone has been reported to have little function-blocking activity, it can augment the inhibitory effects of mAb R1-2 (Cat. No. 553153), resulting in almost complete inhibition of VLA-4 binding to VCAM-1.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49d
Applications Flow Cytometry
Classification Monoclonal
Clone 9C10 (MRF4.B)
Concentration 0.2mg/mL
Conjugate PE
Description Integrin α4; Itga4; Integrin alpha-4; LPAM alpha; VLA-4 alpha; VLA-4a
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse (C57BL/6xA/J)F[1] Fetal Liver Mast MC/9
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.