Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49e Mouse anti-Human, Alexa Fluor 647, Clone: IIA1, BD

Catalog No. BDB563578 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563578 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563578 Supplier BD Biosciences Supplier No. 563578
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The IIA1 monoclonal antibody specifically recognizes CD49e, a type I transmembrane glycoprotein which is also known as Integrin α5 Chain or VLA-5 α chain (VLA-5 α ). Following translation, CD49e is cleaved into two disulfide-linked chains of 135 and 25 kDa that associate with β1 integrin (CD29) to form the VLA-5 (Integrin α5/β1) complex. The VLA-5 complex is a well-established fibronectin receptor that is expressed on many cell types, including fibroblasts, endothelial cells, epithelial cells, platelets, thymocytes, T cells, B cells, the myeloid cell line U937, K562 and some melanoma cell lines. This clone inhibits α5β1 binding of ligands. It is useful for studies of the expression and function of this integrin.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49e
Applications Flow Cytometry
Classification Monoclonal
Clone IIA1
Conjugate Alexa Fluor 647
Description Integrin α5; Integrin alpha-5; ITGA5; ITA5; FNRA; VLA-5; VLA5A
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen P-selectin-IgG1 fusion protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.