Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49e Rat anti-Mouse, Unlabeled, Clone: 5H10-27, BD

Catalog No. BDB553319 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553319 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553319 Supplier BD Biosciences Supplier No. 553319
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 5H10-27 (MFR5) antibody reacts with the α; 5 chain of the integrin α;5 β; 1 fibronectin receptor (CD49e/CD29, VLA-5) on thymocytes, activated T lymphocytes, mast cells, and a variety of mouse cell lines, but not splenocytes. Soluble 5H10-27 (MFR5) antibody has been reported to inhibit VLA-5–mediated functions in vitro. In addition, immobilized mAb 5H10-27 (MFR5) has been demonstrated to costimulate the proliferative response of CD8+ T cells to plate-bound anti-CD3e antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49e
Applications Flow Cytometry
Classification Monoclonal
Clone 5H10-27
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Integrin α5 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse mast cell line MC/9
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.