Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49e Mouse anti-Human, NA/LE, Unlabeled, Clone: IIA1, BD

Catalog No. BDB555614 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555614 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555614 Supplier BD Biosciences Supplier No. 555614
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the α5 integrin, a dimer of 135/25kDa, that associates with β1 integrin (CD29) to form VLA-5, a well-established fibronectin receptor that is expressed on many cell types, including fibroblasts, endothelial cells, epithelial cells, platelets, peripheral T cells, the myeloid cell line U937, K562 and some melanoma cell lines. This clone inhibits α5β1/ligand binding, and is useful for studies of the expression and function of this integrin. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49e
Applications Flow Cytometry
Classification Monoclonal
Clone IIA1
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Integrin α5 Chain
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Immunogen Human α5β1 Protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.