Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD49f Mouse anti-Human, Clone: 4F10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma CBL458
Description
This Anti-CD49f Antibody, clone 4F10 is validated for use in flow cytometry, immunoprecipitation, immunohistochemistry for the detection of CD49f.
Specifications
CD49f | |
Monoclonal | |
Unconjugated | |
NM_000210.2; NM_001079818.1 | |
Mouse | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
4F10 | |
The monoclonal was purified by protein A affinity chromatography and is presented as a liquid in phosphate buffered saline containing 10mM sodium azide and 1mg/mL bovine serum albumin. | |
ITGA6, VLA-6, ITGA6B, CD49f | |
Cell line SW/222 (Human colorectal carcinoma) | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction