Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49f Mouse anti-Human, FITC, Clone: 4F10, MilliporeSigma™

Catalog No. CBL458FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL458FMI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL458FMI Supplier MilliporeSigma Supplier No. CBL458F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD49f Antibody, FITC conjugated, clone 4F10, FITC conjugated is an antibody against CD49fd for use in flow cytometry, immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49f
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 4F10
Conjugate FITC
Formulation Protein A purified immunoglobulin conjugated to fluorescein isothiocyanate (FITC). Liquid in PBS containing 1mg/mL BSA and 10mM sodium azide.
Gene Accession No. NM_000210.2; NM_001079818.1
Gene Symbols ITGA6, VLA-6, ITGA6B, CD49f
Host Species Mouse
Immunogen Cell line SW/222 (Human colorectal carcinoma).
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.