Learn More
Description
Specifications
Specifications
| Antigen | CD49f |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4F10 |
| Conjugate | PE |
| Formulation | The conjugate is supplied in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. For flow cytometry we recommend using 10μL of conjugate per test. |
| Gene Accession No. | NM_000210.2; NM_001079818.1 |
| Gene Symbols | ITGA6, VLA-6, ITGA6B, CD49f |
| Host Species | Mouse |
| Immunogen | Cell line SW/222 (Human colorectal carcinoma) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.