Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4a Mouse, PE, Clone: 74-12-4, BD

Catalog No. BDB559586 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559586 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559586 Supplier BD Biosciences Supplier No. 559586
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 74-12-4 (also known as clone PT4) monoclonal antibody specifically binds to CD4, a 55-kDa antigen expressed on T lymphocytes. This antibody does not react with CTL effectors, CTL precursors, or NK cells (ie, CD8[bright] cells) and it does not cross-react with human or bovine cells. Two peripheral T-helper lymphocyte phenotypes can be distinguished in the pig: CD4+CD8- and CD4+CD8[dull]. mAb 74-12-4 has been reported to inhibit proliferative responses of peripheral blood lymphocytes to mitogen, soluble antigen, and alloantigen. It is only marginally effective for in vivo depletion of peripheral CD4+ T cells. Two alloantigenic forms of CD4 have been recognized in miniature swine based upon their recognition (CD4.1) or lack of recognition (CD4.2) by mAb 74-12-4; the CD4.2 phenotype displays an autosomal recessive, non-MHC-linked, pattern of inheritance. The molecular basis for the polymorphism is a cluster of nucleotide differences leading to multiple amino-acid substitutions in the Ig CDR2-like loop structure. This mAb was clustered as anti-CD4a at the First International Swine CD Workshop. It has been reported to crossreact with chicken leukocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4a
Applications Flow Cytometry
Classification Monoclonal
Clone 74-12-4
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen dd miniature swine thymocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Chicken, Pig
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.