Learn More
CD5 Mouse anti-Human, Biotin, Clone: L17F12, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 347301
Description
CD5, clone L17F12, is derived from hybridization of mouse NS-1/Ag4 myeloma cells with spleen cells from BALB/c mice immunized with human T-acute lymphoblastic leukemia (ALL) cells.
CD5 recognizes a human T-lymphocyte antigen, 67 kilodaltons (kDa).

Specifications
CD5 | |
Monoclonal | |
5μg/mL | |
Phosphate buffered saline with 0.1% sodium azide. | |
Human T-acute lymphoblastic leukemia (ALL) cells | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
L17F12 | |
Biotin | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.