Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Mouse anti-Feline, Lion, R-PE, Clone: f43, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 810009
Description
SouthernBiotech offers anti-feline primary antibodies which are suitable flow cytometry and microscopy applications.
Specifications
CD5 | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Feline T cells | |
RUO | |
Feline, Lion | |
IgG1 |
Flow Cytometry | |
f43 | |
R-PE | |
Mouse | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction