Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Rat anti-Mouse, FITC, Clone: 53-7.3, BD

Catalog No. BDB553020 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553020 0.1 mg
BDB553021 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553020 Supplier BD Biosciences Supplier No. 553020
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 53-7.3 clone has been reported to react with a monomorphic epitope of CD5, a member of the scavenger receptor cysteine-rich protein superfamily and the major ligand of CD72, found on thymocytes, T lymphocytes, thymic NK-T cells, and a subset of B lymphocytes, but not on NK cells or splenic NK-T cells. The level of surface CD5 expression is developmentally regulated in the thymus, starting with low levels on CD4-CD8- thymocytes and increasing as they mature to CD4+CD8+ then CD4+CD8- or CD4-CD8+ thymocytes. Relatively high levels are maintained on peripheral T lymphocytes. The level of CD5 antigen detected on T helper cells has been reported to be somewhat higher than that on T cytotoxic/suppressor and B cells. Few, if any, intestinal intraepithelial lymphocytes bearing the γδ T-cell receptor express CD5. Phenotypic, anatomical, functional, developmental, and pathogenic characteristics of peripheral CD5+ B cells suggest that they may represent a distinct lineage, known as B-1 cells. The frequency of these CD5+ B cells has been reported to show strain-dependent variation. An additional population of CD5+ B lymphocytes resides in the thymus, where it matures from intrathymic B-cell progenitors. It has been proposed that CD5 is a costimulatory molecule which mediates interactions of cells in the immune system and negatively regulates signal transduction mediated by the T-cell receptor and B-cell receptor. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 53-7.3
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Thymus / Spleen

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD5
Applications Flow Cytometry
Classification Monoclonal
Clone 53-7.3
Concentration 0.5mg/mL
Conjugate FITC
Description Lymphocyte antigen 1; Cd5; Ly-12; Ly-A; Lyt-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Thymus / Spleen
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.