Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Mouse anti-Rat, FITC, Clone: OX-19, BD

Catalog No. BDB551449 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551449 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551449 Supplier BD Biosciences Supplier No. 551449
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OX-19 antibody reacts with CD5, a 69 kDa cell-surface glycoprotein found on thymocytes, peripheral T lymphocytes, and some thymic dendritic cells, but not on alloantigen-activated cytotoxic T cells, NK cells, γδ TCR-bearing intestinal intraepithelial lymphocytes, or dendritic epidermal T cells. A CD5+ subset of B lymphocytes has not been characterized in the rat. In the mouse and human, CD72 is the major ligand for CD5. While the OX-19 antibody is not mitogenic, its presence augments in vitro proliferative responses of T cells to lectins and allogeneic cells.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD5
Applications Flow Cytometry
Classification Monoclonal
Clone OX-19
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Thymocyte Lentil Lectin-binding Glycoproteins
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.