Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Mouse anti-Human, PerCP-Cy5.5, Clone: L17F12, BD

Catalog No. BDB341089 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB341089 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB341089 Supplier BD Biosciences Supplier No. 341089
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD5, clone L17F12, is derived from hybridization of mouse NS-1/Ag4 myeloma cells with spleen cells from BALB/c mice immunized with human T-acute lymphoblastic leukemia (ALL) cells.

CD5 recognizes a human T-lymphocyte antigen, 67 kilodaltons (kDa).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD5
Applications Flow Cytometry
Classification Monoclonal
Clone L17F12
Concentration 3μg/mL
Conjugate PerCP-Cyanine5.5
Formulation Phosphate buffered saline with 0.1% sodium azide.
Host Species Mouse
Immunogen Human T-acute lymphoblastic leukemia (ALL) cells
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.