Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-3 Monoclonal Antibody (ICO-60), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA17656
Description
MA1-7656 detects ICAM-3 from human samples. MA1-7656 has been successfully used in flow cytometry procedures. The MA1-7656 immunogen is Leukocytes.
ICAM-3 Monoclonal antibody specifically detects ICAM-3 in Human samples. It is validated for Flow Cytometry.Specifications
ICAM-3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.2% gelatin, 0.5% saccharose and 0.01% sodium azide; pH 7.2 | |
P32942 | |
ICAM3 | |
Leukocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
ICO-60 | |
Unconjugated | |
ICAM3 | |
CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3 | |
Mouse | |
Protein A | |
RUO | |
3385 | |
4°C short term, -80°C long term | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction