Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD51/CD61 Mouse anti-Human, FITC, Clone: 23C6, BD

Catalog No. BDB555505 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555505 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555505 Supplier BD Biosciences Supplier No. 555505
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 23C6 monoclonal antibody specifically binds to the Integrin αvβ3 heterodimeric complex (CD51/CD61) that is expressed on osteoclasts, endothelial cells, macrophages, melanoma cells, some B cells and in very low amounts on platelets. This antibody has been reported to crossreact with chicken and rabbit, but not with rat αvβ3. The integrin αvβ3 heterodimer, often referred to as the vitronectin receptor, binds several ligands in addition to vitronectin including, von Willebrand factor, thrombospondin, osteopontin, bone sialoprotein, neural adhesion molecule L1, laminin, fibronectin, and fibrinogen. The binding of some ligands to the vitronectin receptor has been reported to be inhibited by the 23C6 monoclonal antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD51/CD61
Applications Flow Cytometry
Classification Monoclonal
Clone 23C6
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Bovine, Human, Rabbit
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.