Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD51 Mouse, Unlabeled, Clone: 21, BD

Catalog No. BDB611012 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611012 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611012 Supplier BD Biosciences Supplier No. 611012
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrins are a family of proteins that mediate intercellular adhesion or adherence to extracellular matrix proteins. Their roles are essential for embryonic development, tumor metastasis, organ function, and proper immune cell function. Family members are heterodimers that contain a larger α subunit that is unique to each individual receptor and a smaller β subunit that can be shared by several receptors. Based on β subunit content, integrins are divided into the subfamilies β1, β2, and β3. The β3 subfamily contains the vitronectin receptor (αVβ3) and the platelet protein gpIIb/IIIa. The α chain of the vitronectin receptor (VNRα, CD51, integrin αV) has been reported to consist of a disulfide-linked large extracellular (125kDa) subunit and a smaller (25kDa) membrane-anchored subunit. The large subunit contains multiple sequences with homology to calcium-binding sites in other proteins and an RGD-dependent ligand-binding site, while the small subunit contains a transmembrane domain and a short cytoplasmic domain. VNRα has been reported to be noncovalently associated with the VNR β3-chain (CD61, gpIIIa). The VNR mediates cell adhesion to RGD-containing ligands such as vitronectin, von Willebrand factor, fibrinogen, and thrombospondin.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD51
Applications Western Blot
Classification Monoclonal
Clone 21
Concentration 250μg/mL
Conjugate Unconjugated
Description Integrin αV; Vitronectin Receptor α; VNRα
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD51/VNRα aa. 609-722
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.