Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD51 Rat anti-Mouse, PE, Clone: RMV-7, BD

Catalog No. BDB551187 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551187 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551187 Supplier BD Biosciences Supplier No. 551187
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The rat anti-mouse CD51 (clone RMV-7) antibody reacts with the 140 kDa integrin αV chain. Heterodimers of CD51 with several integrin β chains function as receptors for extracellular matrix proteins. CD51/CD61 (αVβ3 integrin, vitronectin receptor) mediates adhesion to fibronectin, fibrinogen, vitronectin, thrombospondin, von Willebrand factor, and CD31 (PECAM-1). It has been reported to be expressed on activated T lymphocytes, polymorphonuclear granulocytes, blastocysts, and osteoclasts. CD51 has reportedly been found to be undetectable on mouse platelets using either antibody clones H9.2B8 or RMV-7. CD51 also forms heterodimers with CD29 (integrin β1), integrins β5, β

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD51
Applications Flow Cytometry
Classification Monoclonal
Clone RMV-7
Concentration 0.2mg/mL
Conjugate PE
Description Integrin αV chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse (BALB/c) IL-2-activated killer (LAK) Cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.