Learn More
CD52 Mouse anti-Human, PE, Clone: 4C8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562945
Description
The 4C8 monoclonal antibody specifically binds to CD52 which is also known as Cambridge pathology 1 antigen (CAMPATH-1) or Human epididymis-specific protein 5 (HE5). CD52 is a highly N-glycosylated, 25-29 kDa protein whose C-terminus is glycophosphatidylinositol anchored in the membrane. It is highly expressed on the surface of thymocytes and mature lymphocytes but not on their stem cell precursors. It is also expressed on monocytes, dendritic cells, eosinophils and epithelial cells of the epididymis and seminal vesicles but not on neutrophils, plasma cells, platelets or erythrocytes. Although its functional role is not well characterized, the CD52 antigen serves as an exquisitely sensitive target antigen for antibody and complement-mediated lysis of CD52-positive cells. Anti-CD52 antibodies are being used clinically to remove lymphocytes from transplanted bone marrow cell preparations and in the treatment of some malignant diseases.

Specifications
CD52 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Human T Lymphocytes | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
4C8 | |
Cambridge pathology 1 Ag; CAMPATH-1; Epididymal secretory protein E5; HE5 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG3 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.