Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52 Mouse anti-Human, FITC, Clone: HI186, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA182162
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Human CD52 is a glycosylphosphatidylinositol (GPI) anchored antigen expression in a number of different tissues.
Specifications
CD52 | |
Monoclonal | |
0.1 mg/mL | |
CD52 | |
CDW52, CAMPATH-1 antigen, CD52 antigen, cambridge pathology 1 antigen, epididymal secretory protein E5, he5, human epididymis-specific protein 5 | |
Mouse | |
Protein G | |
RUO | |
1043 | |
4°C or -20°C if preferred, store in dark | |
Liquid |
Flow Cytometry | |
HI186 | |
FITC | |
P31358 | |
CD52 | |
Proprietary Immunogen. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction