Learn More
CD53 molecule, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CD53 protein.
Supplier: Abnova Corporation H00000963B01
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins. It contributes to the transduction of CD2-generated signals in T cells and natural killer cells and has been suggested to play a role in growth regulation. Familial deficiency of this gene has been linked to an immunodeficiency associated with recurrent infectious diseases caused by bacteria, fungi and viruses. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq
Sequence: MGMSSLKLLKYVLFFFNLLFWICGCCILGFGIYLLIHNNFGVLFHNLPSLTLGNVFVIVGSIIMVVAFLGCMGSIKENKCLLMSFFILLLIILLAEVTLAILLFVYEQKLNEYVAKGLTDSIHRYHSDNSTKAAWDSIQSFLQCCGINGTSDWTSGPPASCPSDRKVEGCYAKARLWFHSNFLYIGIITICVCVIEVLGMSFALTLNCQIDKTSQTIGLSpecifications
CD53 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human CD53 protein. | |
CD53 | |
MOX44/TSPAN25 | |
Mouse | |
50 μL | |
Yes | |
963 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_000560.3 | |
CD53 | |
CD53 (NP_000551.1, 1 a.a. ∼ 219 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.