Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 Mouse anti-Human, BB515, Clone: HA58, BD

Catalog No. BDB564685 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564685 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564685 Supplier BD Biosciences Supplier No. 564685
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HA58 monoclonal antibody specifically binds to CD54 which is also known as ICAM-1 (intracellular adhesion molecule-1). CD54 is an 85-110 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin supergene family. A soluble form of CD54 can also be found in biological fluids. CD54 is expressed on endothelial cells and both resting (weak) and activated (moderate) lymphocytes and monocytes. CD54 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). The CD54 adhesion molecule plays roles in inflammatory and immune responses and neoplasia. This antibody (NA/LE format) blocks the mixed-lymphocyte reaction (MLR) and the purified format is suitable for staining acetone-fixed, frozen tissue sections.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54
Applications Flow Cytometry
Classification Monoclonal
Clone HA58
Conjugate Brilliant Blue 515 (BB515)
Description ICAM-1; intercellular adhesion molecule 1; BB2; P3.58
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen IFN-γ-treated BM314 colonic carcinoma cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.