Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 Mouse anti-Rat, Unlabeled, Clone: 1A29, BD

Catalog No. BDB554967 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554967 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554967 Supplier BD Biosciences Supplier No. 554967
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1A29 antibody reacts with ICAM-1 (CD54), an 85kDa surface glycoprotein expressed on vascular endothelium in lymphoid tissues, thymic stromal cells, peripheral blood monocytes, peritoneal macrophages and mast cells, dendritic cells, and weakly on peripheral lymphocytes and thymocytes. ICAM-1 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). Its expression is upregulated on activated lymphocytes and endothelial cells. 1A29 mAb inhibits PMA-induced aggregation of PHA-stimulated splenic blasts, as well as the adhesion of mitogen-stimulated blasts to HEV-derived endothelial cells and purified ICAM-1. It has also been reported that 1A29 antibody can inhibit leukocyte infiltration in in vivo systems, blocks induction of experimental allergic encephalomyelitis, and reduces NK-cell adhesion to tumor cells. The effects of in vivo administration of the 1A29 monoclonal antibody have been thoroughly investigated.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54
Applications Flow Cytometry
Classification Monoclonal
Clone 1A29
Concentration 0.5mg/mL
Conjugate Unconjugated
Description ICAM-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat HEV-Derived Cell Line Ax
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.