Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 Mouse anti-Human, Unlabeled, Clone: HA58, BD

Catalog No. BDB555510 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555510 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555510 Supplier BD Biosciences Supplier No. 555510
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the 85-110kDa integral membrane glycoprotein, also known as intracellular adhesion molecule-1 (ICAM-1), expressed on endothelial cells and both resting (weak) and activated (moderate) lymphocytes and monocytes. CD54 is a ligand for the leukocyte function antigen-1 (CD11a). CD54 antibodies are used for the studies of inflammatory processes and neoplasia. This antibody (NA/LE format) blocks the mixed-lymphocyte reaction (MLR) and the purified format is suitable for staining acetone-fixed, frozen tissue sections. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54
Applications Flow Cytometry
Classification Monoclonal
Clone HA58
Concentration 0.5mg/mL
Conjugate Unconjugated
Description ICAM-1; intercellular adhesion molecule 1; BB2; P3.58
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen IFN-γ-treated BM314 colonic carcinoma cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.