Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 (ICAM-1) Mouse anti-Human, APC, Clone: 15.2, MilliporeSigma™

Catalog No. MABF711MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF711MI 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF711MI Supplier MilliporeSigma Supplier No. MABF711
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal antibody Anti-CD54 (ICAM-1) (human), APC, clone 15.2 validated for use in western blotting, immunohistochemistry and Flow Cytometry for the detection of CD54 (ICAM-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54 (ICAM-1)
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 15.2
Concentration 5μL (0.5μg)/test
Conjugate APC
Formulation Purified Mouse Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols ICAM1
Host Species Mouse
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 25 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.