Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 (Leu -54) Mouse anti-Human, PE, Clone: LB-2, BD

Catalog No. BDB347977 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347977 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347977 Supplier BD Biosciences Supplier No. 347977
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD54, clone LB-2, is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with the major histocompatibility complex (MHC) class II–negative human B-lymphoblastoid cell line 6.16c13.

CD54 (Leu-54) recognizes the intercellular adhesion molecule (ICAM-1), Mr 90 kdaltons (kDa). The CD54 antigen is a ligand for the leucocyte function–associated antigen-1 (CD11a/CD18 [LFA-1]) and influences both LFA-1–dependent adhesion of leucocytes to endothelial cells and immune functions involving cell-to-cell contact.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54 (Leu -54)
Applications Flow Cytometry
Classification Monoclonal
Clone LB-2
Concentration 3μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Major histocompatibility Class II antigen negative variant of human B-lymphoblastoid cell line
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.