Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD55 Mouse anti-Human, Clone: BRIC 216, MilliporeSigma™

Catalog No. CBL511MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL511MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL511MI Supplier MilliporeSigma Supplier No. CBL511
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD55 Antibody, clone BRIC 216 is an antibody against CD55 for use in flow cytometry, immunoprecipitation, western blot, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD55
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BRIC 216
Conjugate Unconjugated
Formulation The immunoglobulin was purified from cell culture supernatant by affinity chromatography using protein G and is presented in 1MTris-buffered saline, pH 7.5, containing no preservatives. The antibody has been sterile-filtered.
Gene Accession No. NM_000574.2
Gene Symbols CD55, CR, TC, DAF
Host Species Mouse
Immunogen Human fibroblast cell line
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.