Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD56 Mouse anti-Human, Unlabeled, Clone: MY31, BD

Catalog No. BDB347740 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347740 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347740 Supplier BD Biosciences Supplier No. 347740
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD56, clone MY31, is derived from the hybridization of mouse Sp2/0 myeloma cells with spleen cells from (B6 x BALB/c) F1 mice immunized with the KG1a cell line.

CD56 recognizes an antigen present on natural killer (NK) lymphocytes that is the 140-kdalton (kDa) isoform of the neural cell adhesion molecule (NCAM). The 140-kDa core protein is extensively glycosylated to give a mature antigen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD56
Applications Flow Cytometry
Classification Monoclonal
Clone MY31
Concentration 50μg/mL
Conjugate Unconjugated
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.