Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD56 Mouse anti-Human, Unlabeled, Clone: MY31, BD

Catalog No. BDB559049 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559049 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559049 Supplier BD Biosciences Supplier No. 559049
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the neural cell adhesion molecule (N-CAM), CD56 antigen, 175-180 kD, a glycoprotein on natural killer (NK) lymphocytes, a subset of T lymphocytes and interleukin-2 (IL-2)-activated thymocytes, as well as neural and degenerating or diseased muscle tissue. Anti-N-CAM monoclonal antibody (clone MY31) immunoprecipitates NCAM from neuroblastoma, KG-1a.5, N-CAM-trasfected mouse L cells, and human adenocarcinoma cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD56
Applications Flow Cytometry
Classification Monoclonal
Clone MY31
Concentration 1.0mg/mL
Conjugate Unconjugated
Description N-CAM
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.