Learn More
CD56 (MY31) PE Mouse anti-Human, PE, Clone: MY31, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 347747
Description
CD56, clone MY31, is derived from the hybridization of mouse Sp2/0 myeloma cells with spleen cells from (B6 x BALB/c) F1 mice immunized with the KG1a cell line.
CD56 recognizes an antigen present on natural killer (NK) lymphocytes that is the 140-kdalton (kDa) isoform of the neural cell adhesion molecule (NCAM). The 140-kDa core protein is extensively glycosylated to give a mature antigen.
Specifications
| CD56 | |
| Monoclonal | |
| 50μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| 100 Tests | |
| Immunology | |
| Human | |
| Purified |
| Flow Cytometry | |
| MY31 | |
| PE | |
| Mouse | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.