Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD56 (N-CAM) Mouse anti-Human, APC, Clone: NCAM16.2, BD

Catalog No. BDB341025 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB341025 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB341025 Supplier BD Biosciences Supplier No. 341025
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD56 antibody, clone NCAM16.2, is derived from the hybridization of P3-X63-Ag8.653 mouse myeloma cells with spleen cells isolated from BALB/c mice immunized with immunoaffinity-enriched NCAM from detergent extracts of adult human brain.

The CD56 antibody recognizes a heavily glycosylated 140-kilodalton (kDa) isoform of NCAM, a member of the immunoglobulin (Ig) superfamily. The CD56 antibody also recognizes 180-kDa and 120-kDa isoforms of NCAM found in neurons and muscle, respectively.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD56
Applications Flow Cytometry
Classification Monoclonal
Clone NCAM16.2
Concentration 12μg/mL
Conjugate APC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Immunoaffinity-enriched adult human brain NCAM
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.