Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD57 Mouse anti-Human, Unlabeled, Clone: HNK-1, BD

Catalog No. BDB559048 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559048 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559048 Supplier BD Biosciences Supplier No. 559048
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HNK-1 monoclonal antibody recognizes a 110 kD glycoprotein (CD57) that is expressed on a subset of natural killer (NK) lymphocytes and T lymphocytes. It also recognizes epitopes on neural or neuroectodermal tumors including neurofibromas, malignant melanomas, malignant peripheral neuroectodermal tumors, Ewing's sarcomas, and small-cell lung carcinomas. Anti-HNK-1 monoclonal antibody also recognizes a carbohydrate epitope on subpopulations of several cell-adhesion molecules including N-CAM and myelin-associated glycoprotein (MAG).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD57
Applications Flow Cytometry
Classification Monoclonal
Clone HNK-1
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Membrane extracts of the HSB-2 T-lymphoblastoid cell line
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.