Learn More
CD58 Mouse anti-Human, Unlabeled, Clone: 1C3, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555919
Description
The 1C3 (AICD58.6) monoclonal antibody specifically binds to CD58. CD58 is a a 60-70 kDa glycoprotein member of the immunoglobulin superfamily. CD58, also referred to as the lymphocyte function-associated antigen-3 (LFA-3), has a wide tissue distribution, being expressed on both hematopoietic and non-hematopoietic cells, including endothelial cells and fibroblasts. There are two isoforms of CD58: a glycosylphosphatidylinositol (GPI)-linked form and a transmembrane form. Both isoforms may be expressed on the same cell type. Erythrocytes, however, only express the GPI-linked isoform. CD58 interacts with CD2 during cell adhesion. This binding can enhance antigen-specific T-cell activation. This interaction can also play a role in cell-mediated cytotoxicity.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD58 | |
Monoclonal | |
0.5mg/mL | |
LFA-3; LFA3; Lymphocyte function-associated antigen 3; Ag3 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
1C3 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Human CD58 | |
0.1 mg | |
Immunology | |
Canine, Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.