Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD58 Mouse anti-Human, FITC, Clone: 1C3, BD

Catalog No. BDB555920 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555920 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555920 Supplier BD Biosciences Supplier No. 555920
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1C3 (AICD58.6) monoclonal antibody specifically binds to CD58. CD58 is a a 60-70 kDa glycoprotein member of the immunoglobulin superfamily. CD58, also referred to as the lymphocyte function-associated antigen-3 (LFA-3), has a wide tissue distribution, being expressed on both hematopoietic and non-hematopoietic cells, including endothelial cells and fibroblasts. There are two isoforms of CD58: a glycosylphosphatidylinositol (GPI)-linked form and a transmembrane form. Both isoforms may be expressed on the same cell type. Erythrocytes, however, only express the GPI-linked isoform. CD58 interacts with CD2 during cell adhesion. This binding can enhance antigen-specific T-cell activation. This interaction can also play a role in cell-mediated cytotoxicity.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD58
Applications Flow Cytometry
Classification Monoclonal
Clone 1C3
Conjugate FITC
Description LFA-3; LFA3; Lymphocyte function-associated antigen 3; Ag3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human CD58
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.