Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD58 (LFA-3) Mouse anti-Human, PE, Clone: L306.4, BD

Catalog No. BDB340295 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340295 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340295 Supplier BD Biosciences Supplier No. 340295
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD58 (Anti-LFA-3), clone L306.4, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from C3H/HeN mice immunized with Lc-LFA-3 mouse L-cell transfectant.

CD58 (Anti-LFA-3) recognizes a 40 to 65-kdalton cell-surface glycoprotein that is a member of the immunoglobulin superfamily. The CD58 antigen, known as the lymphocyte function–associated antigen-3 (LFA-3), mediates cellular adhesion and participates in signal transduction when it binds to its ligand, the CD2 antigen. Cellular interactions regulated by the CD58/CD2 antigens are involved in the antigen-independent adhesion pathway and cytotoxic T lymphocyte (CTL) activity. The CD58 antigen has two isoforms. One isoform is anchored in the cell membrane by a glycophosphatidyl inositol tail, while the other has a transmembrane hydrophobic segment and a cytoplasmic segment composed of 12 amino acids.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD58
Applications Flow Cytometry
Classification Monoclonal
Clone L306.4
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Lc-LFA-3 mouse L-cell transfectant
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.